COMH4211B1C8 Combi-Blocks, Inc.

2-Methoxyethyl (4-nitrophenyl) carbonate

  • MDL number MFCD26395377
  • Cas Number : 426264-10-0
  • Molecular Weight : 241.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4070

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4070
Availability details for
Quantity
  • Availability
  • Add to Cart