COMH4211B1CA Combi-Blocks, Inc.

(4-Cyclopropylmethoxy-pyrazol-1-yl)-acetic acid

  • MDL number MFCD31579320
  • Cas Number : 2270906-19-7
  • Molecular Weight : 196.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4080

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4080
Availability details for
Quantity
  • Availability
  • Add to Cart