COMH4211B1CC Combi-Blocks, Inc.

5-(4-Bromophenyl)-2-cyclopropyl-1h-imidazole

  • MDL number MFCD20260003
  • Cas Number : 1242441-13-9
  • Molecular Weight : 263.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4087

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4087
Availability details for
Quantity
  • Availability
  • Add to Cart