COMH4211B1CE Combi-Blocks, Inc.

4-[(2R,6S)-2,6-Dimethylmorpholin-4-yl]benzaldehyde

  • MDL number MFCD31579439
  • Cas Number : 736991-27-8
  • Molecular Weight : 219.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4091

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4091
Availability details for
Quantity
  • Availability
  • Add to Cart