COMH4211B1CF Combi-Blocks, Inc.

1-[(2-Fluorophenyl)methyl]-1h-pyrazol-4-ol

  • MDL number MFCD29918910
  • Cas Number : 1599923-23-5
  • Molecular Weight : 192.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4092

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4092
Availability details for
Quantity
  • Availability
  • Add to Cart