COMH4211B1D1 Combi-Blocks, Inc.

6-Chloro-n,n'-dicyclopropyl-[1,3,5]triazine-2,4-diamine

  • MDL number MFCD30492336
  • Cas Number : 22936-82-9
  • Molecular Weight : 225.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4094

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4094
Availability details for
Quantity
  • Availability
  • Add to Cart