COMH4211B1D2 Combi-Blocks, Inc.

Ethyl 2-(4-cyano-1h-pyrazol-1-yl)acetate

  • MDL number MFCD29058438
  • Cas Number : 1630080-88-4
  • Molecular Weight : 179.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4097

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4097
Availability details for
Quantity
  • Availability
  • Add to Cart