COMH4211B1D4 Combi-Blocks, Inc.

2-Chloro-3-(1-fluorocyclobutyl)-6-methylpyridine

  • MDL number MFCD31613718
  • Cas Number : 1613292-61-7
  • Molecular Weight : 199.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4101

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4101
Availability details for
Quantity
  • Availability
  • Add to Cart