COMH4211B1D5 Combi-Blocks, Inc.

1,4-Dibromo-2-fluorobutane

  • MDL number MFCD20482701
  • Cas Number : 112464-88-7
  • Molecular Weight : 233.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4102

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4102
Availability details for
Quantity
  • Availability
  • Add to Cart