COMH4211B1D7 Combi-Blocks, Inc.

N-(Cyclopropylmethyl)-4-iodobenzamide

  • MDL number MFCD14647620
  • Cas Number : 39887-31-5
  • Molecular Weight : 301.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4110

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4110
Availability details for
Quantity
  • Availability
  • Add to Cart