COMH4211B1D8 Combi-Blocks, Inc.

3-(1-Ethyl-piperidin-4-yl)-propionic acid hydrochloride

  • MDL number MFCD31557821
  • Cas Number : 1087728-65-1
  • Molecular Weight : 221.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4114

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4114
Availability details for
Quantity
  • Availability
  • Add to Cart