COMH4211B1D9 Combi-Blocks, Inc.

((3-Bromopropoxy)methanetriyl)tribenzene

  • MDL number MFCD31618936
  • Cas Number : 76504-33-1
  • Molecular Weight : 381.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4118

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4118
Availability details for
Quantity
  • Availability
  • Add to Cart