COMH4211B1DA Combi-Blocks, Inc.

4-(2-Fluoro-4-methoxy-phenyl)-piperidine hydrochloride

  • MDL number MFCD31583397
  • Cas Number : 2270913-21-6
  • Molecular Weight : 245.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4119

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4119
Availability details for
Quantity
  • Availability
  • Add to Cart