COMH4211B1DD Combi-Blocks, Inc.

Methyl 5-chloro-4-iodo-2-methoxybenzoate

  • MDL number MFCD23716231
  • Cas Number : 473574-26-4
  • Molecular Weight : 326.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4121

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4121
Availability details for
Quantity
  • Availability
  • Add to Cart