COMH4211B1E3 Combi-Blocks, Inc.

[4-Bromo-2-(trifluoromethyl)phenyl]hydrazine

  • MDL number MFCD02663116
  • Cas Number : 312303-71-2
  • Molecular Weight : 255.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-[4-bromo-2-(trifluoromethyl)phenyl]hydrazine
Inchi Code 1S/C7H6BrF3N2/c8-4-1-2-6(13-12)5(3-4)7(9,10)11/h1-3,13H,12H2
Inchi Key XGLDQVFQOXGKKS-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4142

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4142
Availability details for
Quantity
  • Availability
  • Add to Cart