COMH4211B1E4 Combi-Blocks, Inc.

4-Amino-n,n-dimethylbutanamide

  • MDL number MFCD09946311
  • Cas Number : 87639-91-6
  • Molecular Weight : 130.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4144

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4144
Availability details for
Quantity
  • Availability
  • Add to Cart