COMH4211B1E6 Combi-Blocks, Inc.

3-(Cyclohexylmethoxy)propanoic acid

  • MDL number MFCD16807355
  • Cas Number : 914305-97-8
  • Molecular Weight : 186.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(cyclohexylmethoxy)propanoic acid
Inchi Code 1S/C10H18O3/c11-10(12)6-7-13-8-9-4-2-1-3-5-9/h9H,1-8H2,(H,11,12)
Inchi Key GMZSVHMYBSEQHQ-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4152

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4152
Availability details for
Quantity
  • Availability
  • Add to Cart