COMH4211B1E7 Combi-Blocks, Inc.

3-Bromo-4-(4-fluoro-phenylsulfanyl)-benzonitrile

  • MDL number MFCD25270853
  • Cas Number : 1549236-76-1
  • Molecular Weight : 308.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4153

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4153
Availability details for
Quantity
  • Availability
  • Add to Cart