COMH4211B1E9 Combi-Blocks, Inc.

[2-(2-Bromo-4-chlorophenyl)ethoxy](tert-butyl)dimethylsilane

  • MDL number MFCD31614137
  • Cas Number : 1858270-47-9
  • Molecular Weight : 349.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4156

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4156
Availability details for
Quantity
  • Availability
  • Add to Cart