COMH4211B1EA Combi-Blocks, Inc.

2-(3-Methoxy-propoxy)-5-methyl-benzylamine

  • MDL number MFCD18331118
  • Cas Number : 1340507-06-3
  • Molecular Weight : 209.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4158

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4158
Availability details for
Quantity
  • Availability
  • Add to Cart