COMH4211B1ED Combi-Blocks, Inc.

4-Iodo-2-methyl-1-(trifluoromethyl)benzene

  • MDL number MFCD27933385
  • Cas Number : 930599-57-8
  • Molecular Weight : 286.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4169

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4169
Availability details for
Quantity
  • Availability
  • Add to Cart