COMH4211B1F2 Combi-Blocks, Inc.

[4-(3-Bromophenoxy)butoxy]-tert-butyldimethylsilane

  • MDL number MFCD26388517
  • Cas Number : 790696-47-8
  • Molecular Weight : 359.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4183

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4183
Availability details for
Quantity
  • Availability
  • Add to Cart