COMH4211B1F3 Combi-Blocks, Inc.

1-(3-Chloro-4-methylphenyl)propan-1-amine

  • MDL number MFCD18702552
  • Cas Number : 1270440-34-0
  • Molecular Weight : 183.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4184

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4184
Availability details for
Quantity
  • Availability
  • Add to Cart