COMH4211B1F4 Combi-Blocks, Inc.

[4-(2-Fluoro-4-methoxy-phenyl)-piperidin-1-yl]-acetic acid

  • MDL number MFCD31583400
  • Cas Number : 2270905-15-0
  • Molecular Weight : 267.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4185

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4185
Availability details for
Quantity
  • Availability
  • Add to Cart