COMH4211B1F5 Combi-Blocks, Inc.

5-Bromo-2-ethylsulfanyl-nicotinic acid tert-butyl ester

  • MDL number MFCD31557782
  • Cas Number : 2204559-72-6
  • Molecular Weight : 318.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4188

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4188
Availability details for
Quantity
  • Availability
  • Add to Cart