COMH4211B1F6 Combi-Blocks, Inc.

N-(3-Bromo-5-methylphenyl)propane-2-sulfonamide

  • MDL number MFCD27942841
  • Cas Number : 1269233-13-7
  • Molecular Weight : 292.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4189

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4189
Availability details for
Quantity
  • Availability
  • Add to Cart