COMH4211B1F7 Combi-Blocks, Inc.

5-Bromo-2-pyrrolidin-1-yl-nicotinic acid tert-butyl ester

  • MDL number MFCD31557806
  • Cas Number : 2206608-41-3
  • Molecular Weight : 327.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4190

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4190
Availability details for
Quantity
  • Availability
  • Add to Cart