COMH4211B1F9 Combi-Blocks, Inc.

3-[4-(3-Bromo-phenyl)-piperazin-1-yl]-propionic acid dihydrochloride

  • MDL number MFCD31583325
  • Cas Number : 2270906-13-1
  • Molecular Weight : 386.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4194

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4194
Availability details for
Quantity
  • Availability
  • Add to Cart