COMH4211B1FA Combi-Blocks, Inc.

1-(4-Chloro-3-fluoro-benzyl)-1h-pyrazol-4-ol

  • MDL number MFCD31561514
  • Cas Number : 1696437-91-8
  • Molecular Weight : 226.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4196

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4196
Availability details for
Quantity
  • Availability
  • Add to Cart