COMH4211B1FB Combi-Blocks, Inc.

(4-Nitro-3-trifluoromethyl-phenoxy)-acetic acid

  • MDL number MFCD27443949
  • Cas Number : 113405-09-7
  • Molecular Weight : 265.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(4-nitro-3-(trifluoromethyl)phenoxy)acetic acid
Inchi Code 1S/C9H6F3NO5/c10-9(11,12)6-3-5(18-4-8(14)15)1-2-7(6)13(16)17/h1-3H,4H2,(H,14,15)
Inchi Key FMMDNTXLWDHKSS-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4197

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4197
Availability details for
Quantity
  • Availability
  • Add to Cart