COMH4211B1FC Combi-Blocks, Inc.

5-(((tert-Butyldimethylsilyl)oxy)methyl)pyridin-2-amine

  • MDL number MFCD20528021
  • Cas Number : 322691-19-0
  • Molecular Weight : 238.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4198

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4198
Availability details for
Quantity
  • Availability
  • Add to Cart