COMH4211B219 Combi-Blocks, Inc.

5-Bromo-2-isopropylamino-nicotinic acid tert-butyl ester

  • MDL number MFCD31557818
  • Cas Number : 2206608-81-1
  • Molecular Weight : 315.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4201

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4201
Availability details for
Quantity
  • Availability
  • Add to Cart