COMH4211B21B Combi-Blocks, Inc.

2-Bromo-1-tert-butyl-4-chlorobenzene

  • MDL number MFCD28356425
  • Cas Number : 1369776-71-5
  • Molecular Weight : 247.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4204

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4204
Availability details for
Quantity
  • Availability
  • Add to Cart