COMH4211B21C Combi-Blocks, Inc.

3-(6-Bromo-pyridin-3-yl)-acrylic acid ethyl ester

  • MDL number MFCD23378795
  • Cas Number : 656827-74-6
  • Molecular Weight : 256.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4208

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4208
Availability details for
Quantity
  • Availability
  • Add to Cart