COMH4211B21F Combi-Blocks, Inc.

2,5-Pyrrolidinedione, 1-[[(9z,12z)-1-oxo-9,12-octadecadienyl]oxy]-

  • MDL number MFCD26792810
  • Cas Number : 69888-88-6
  • Molecular Weight : 377.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4216

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4216
Availability details for
Quantity
  • Availability
  • Add to Cart