COMH4211B220 Combi-Blocks, Inc.

3-Bromo-n,n,2-trimethylbenzamide

  • MDL number MFCD21252747
  • Cas Number : 1369848-53-2
  • Molecular Weight : 242.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4217

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4217
Availability details for
Quantity
  • Availability
  • Add to Cart