COMH4211B221 Combi-Blocks, Inc.

5-(4-Bromo-2,6-difluorophenyl)-1,3,4-thiadiazol-2-amine

  • MDL number MFCD23840455
  • Cas Number : 1526200-39-4
  • Molecular Weight : 292.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4219

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4219
Availability details for
Quantity
  • Availability
  • Add to Cart