COMH4211B222 Combi-Blocks, Inc.

Benzyl cis-(2-hydroxymethyl)cyclohexylcarbamate

  • MDL number MFCD03844617
  • Cas Number : 213672-66-3
  • Molecular Weight : 263.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4220

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4220
Availability details for
Quantity
  • Availability
  • Add to Cart