COMH4211B223 Combi-Blocks, Inc.

5-Chloro-2-cyclopropylmethylsulfanyl-phenylamine

  • MDL number MFCD12138863
  • Cas Number : 1154384-91-4
  • Molecular Weight : 213.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4222

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4222
Availability details for
Quantity
  • Availability
  • Add to Cart