COMH4211B224 Combi-Blocks, Inc.

5-Bromo-2-(3-fluoro-phenoxy)-nicotinic acid tert-butyl ester

  • MDL number MFCD31557864
  • Cas Number : 2206609-23-4
  • Molecular Weight : 368.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4225

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4225
Availability details for
Quantity
  • Availability
  • Add to Cart