COMH4211B227 Combi-Blocks, Inc.

5-Benzyloxy-2-(2-methoxy-ethoxy)-benzoic acid

  • MDL number MFCD31618915
  • Molecular Weight : 302.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4227

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4227
Availability details for
Quantity
  • Availability
  • Add to Cart