COMH4211B22A Combi-Blocks, Inc.

4-Chloro-2-(4-methoxy-pyrazol-1-ylmethyl)-benzoic acid

  • MDL number MFCD31583340
  • Cas Number : 2270909-18-5
  • Molecular Weight : 266.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4239

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4239
Availability details for
Quantity
  • Availability
  • Add to Cart