COMH4211B22B Combi-Blocks, Inc.

2-Methyl-2-(4-propoxy-pyrazol-1-yl)-propionic acid

  • MDL number MFCD31579341
  • Cas Number : 2270906-89-1
  • Molecular Weight : 212.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4245

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4245
Availability details for
Quantity
  • Availability
  • Add to Cart