COMH4211B22C Combi-Blocks, Inc.

4-(4-Aminophenylthio)pyridin-2-amine

  • MDL number MFCD23131135
  • Cas Number : 1020087-60-8
  • Molecular Weight : 217.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4246

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4246
Availability details for
Quantity
  • Availability
  • Add to Cart