COMH4211B22D Combi-Blocks, Inc.

6-Bromo-2-(4-fluorophenyl)imidazo[1,2-a]pyridine

  • MDL number MFCD06641501
  • Cas Number : 426825-66-3
  • Molecular Weight : 291.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4247

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4247
Availability details for
Quantity
  • Availability
  • Add to Cart