COMH4211B22F Combi-Blocks, Inc.

2-Bromo-1-(cyclopentyloxy)-4-fluorobenzene

  • MDL number MFCD09930381
  • Cas Number : 1016505-33-1
  • Molecular Weight : 259.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4254

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4254
Availability details for
Quantity
  • Availability
  • Add to Cart