COMH4211B231 Combi-Blocks, Inc.

4-(1-Bromonaphthalen-4-yl)phenol

  • MDL number MFCD28965590
  • Cas Number : 1312609-73-6
  • Molecular Weight : 299.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4264

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4264
Availability details for
Quantity
  • Availability
  • Add to Cart