COMH4211B232 Combi-Blocks, Inc.

4-[4-(2,2,2-Trifluoro-ethoxy)-phenyl]-butyric acid

  • MDL number MFCD31561458
  • Cas Number : 2270906-32-4
  • Molecular Weight : 262.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4267

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4267
Availability details for
Quantity
  • Availability
  • Add to Cart