COMH4211B234 Combi-Blocks, Inc.

1-Piperidinecarboxylic acid, 4-[(4-formylphenoxy)methyl]-, 1,1-dimethylethyl ester

  • MDL number MFCD24468176
  • Cas Number : 170726-00-8
  • Molecular Weight : 319.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4271

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4271
Availability details for
Quantity
  • Availability
  • Add to Cart