COMH4211B235 Combi-Blocks, Inc.

5-Bromo-n,2-dimethoxybenzamide

  • MDL number MFCD31628784
  • Cas Number : 1326614-35-0
  • Molecular Weight : 260.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4272

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4272
Availability details for
Quantity
  • Availability
  • Add to Cart